Learn More
Grace™ Alltech™ GracePure™ SPE Columns
Clean up and concentrate problematic samples
Brand:  Grace™ 5141520
 View more versions of this product
Product Code. 10282370
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Extensively tested and tightly controlled media
  • Less than 2% bed weight variation
  • Excellent value
Specifications
Specifications
6 mL
Cleanup, Concentration and Recovery of Chromatography Samples
Solid Phase Extraction Column
1000 mg
Florisil