Learn More
Grace™ Alltech™ GracePure™ SPE Columns
Clean up and concentrate problematic samples
Brand: Grace™ 5138777
This item is not returnable.
View return policy
Description
- Extensively tested and tightly controlled media
- Less than 2% bed weight variation
- Excellent value
Specifications
Silica | |
Normal | |
100 mg | |
50 μm |
1 mL | |
SPE | |
Cleanup, Concentration and Recovery of Chromatography Samples | |
60 Å |