missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poeppelmann™ GPN 985 Series LDPE Grease Nipple Cap
Click to view available options
Type:
Grease Nipple Cap
Grease Nipple Cap with Tab
Color:
Red
Yellow
Unit Size:
Pack of 100
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | Protect the tapered head of grease nipples against contamination and visibly mark lubrication points |
| Length (Metric) | 12 mm |
| Quantity | 100 Pack |
| Diameter (Metric) Outside | 12 mm |
| Material | LDPE |
| Type | Grease Nipple Cap with Tab |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction