Learn More
Poeppelmann™ KAPSTO™ GPN 800 Screw Caps
Features easy-to-grip external rib for easy mounting and removal by hand. Poeppelmann™ KAPSTO™ GPN 800 Screw Caps are manufactured from yellow PE-HD plastic.
Brand:  Poeppelmann™ 80026150000
 View more versions of this product
Product Code. 10768415
  • € 61.40 / Pack of 100
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With metric threads
  • Resistant to alcohol and weak acids/alkalis
  • Suitable for a variety of different thread types
Specifications
Specifications
Yellow
M 26 x 1.5 Threads
100/Pk.
34 mm
Screw Cap
Suitable for a variety of different thread types
13 mm
O.D. x H: 34 x 13 mm, M 26 x 1.5 Thread Type, 11 mm Interior Height
HDPE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers