Learn More
Poeppelmann™ KAPSTO™ GPN 800 Screw Caps
Features easy-to-grip external rib for easy mounting and removal by hand. Poeppelmann™ KAPSTO™ GPN 800 Screw Caps are manufactured from yellow PE-HD plastic.
Brand:  Poeppelmann™ 80069010000
 View more versions of this product
Product Code. 10687605
  • € 111.00 / Pack of 100
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With metric threads
  • Resistant to alcohol and weak acids/alkalis
  • Suitable for a variety of different thread types
Specifications
Specifications
Yellow
G 2 Threads
100/Pk.
68 mm
Screw Cap
Suitable for a variety of different thread types
20 mm
O.D. x H: 68 x 20 mm, G 2 Thread Type, 18 mm Interior Height
HDPE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers