Learn More
Poeppelmann™ KAPSTO™ GPN 800 Screw Caps
Features easy-to-grip external rib for easy mounting and removal by hand. Poeppelmann™ KAPSTO™ GPN 800 Screw Caps are manufactured from yellow PE-HD plastic.
Brand:  Poeppelmann™ 80024150000
 View more versions of this product
Product Code. 10564715
  • € 55.40 / Pack of 100
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With metric threads
  • Resistant to alcohol and weak acids/alkalis
  • Suitable for a variety of different thread types
Specifications
Specifications
Yellow
M 24 x 1.5 Threads
100/Pk.
30 mm
Screw Cap
Suitable for a variety of different thread types
15.5 mm
O.D. x H: 30 x 15.5 mm, M 24 x 1.5 Thread Type
HDPE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers