Learn More
Poeppelmann™ KAPSTO™ GPN 800 Screw Caps
Features easy-to-grip external rib for easy mounting and removal by hand. Poeppelmann™ KAPSTO™ GPN 800 Screw Caps are manufactured from yellow PE-HD plastic.
Brand:  Poeppelmann™ 80036150000
 View more versions of this product
Product Code. 10237714
  • € 73.25 / Pack of 100
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Description
  • With metric threads
  • Resistant to alcohol and weak acids/alkalis
  • Suitable for a variety of different thread types
Specifications
Specifications
Yellow
M 36 x 1.5 Threads
100/Pk.
44 mm
Screw Cap
Suitable for a variety of different thread types
19 mm
O.D. x H: 44 x 19 mm, M 36 x 1.5 Thread Type
HDPE