Learn More
Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs
Features easy-to-grip head for manual assembly. Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs are manufactured from yellow PE-HD plastic.
Brand:  Poeppelmann™ 70000100000
 View more versions of this product
Product Code. 10480255
  • € 25.35 / Pack of 100
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Description
  • With metric threads
  • Acc. with DIN EN ISO 228-1
  • Resistant to alcohol and weak acids/alkalis
  • Small to medium sizes feature a slot for screwdriver installation
  • Suitable for a variety of metric and imperial threads, as well as pipe threads
Specifications
Specifications
14 mm
Screw Plug Closure
Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1
M 10
Yellow
14 mm