Learn More
Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs
Features easy-to-grip head for manual assembly. Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs are manufactured from yellow PE-HD plastic.
Brand:  Poeppelmann™ 70016150000
 View more versions of this product
Product Code. 10073674
  • € 32.15 / Pack of 100
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With metric threads
  • Acc. with DIN EN ISO 228-1
  • Resistant to alcohol and weak acids/alkalis
  • Small to medium sizes feature a slot for screwdriver installation
  • Suitable for a variety of metric and imperial threads, as well as pipe threads
Specifications
Specifications
21 mm
Screw Plug Closure
Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1
M 16 x 1.5
Yellow
16 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers