Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures are manufactured from natural-colored PE-LD plastic.
Brand:  Poeppelmann™ 60011400000
 View more versions of this product
Product Code. 10023514
  • € 118.00 / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Description
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Specifications
Natural
17.2 mm
O.D. x H: 125 x 17.2 mm
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
50 Pack
125 mm
Tapered Closure