missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 17mm Depth
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures are manufactured from natural-colored PE-LD plastic.
Brand: Poeppelmann™ 60007750000
This item is not returnable.
View return policy
Description
- 17mm insertion depth
- Universal use as plug or cap
- Tapered design ideal for accomodating tolerances
- Chemically resistant to alcohol and weak acids/alkalis
Specifications
Natural | |
17.2 mm | |
O.D. x H: 88 x 17.2 mm, 17 mm Insertion Depth | |
LDPE |
For use as plugs in bores or internal thread, or as caps for external thread | |
100 Pack | |
88 mm | |
Tapered Closure |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction