Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 13.5mm Depth
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 13.5mm Depth are manufactured from natural-colored PE-LD plastic.
Brand:  Poeppelmann™ 60005450000
 View more versions of this product
Product Code. 10256844
  • € 47.60 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Description
  • 13.5mm insertion depth
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Specifications
Natural
13.5 mm
O.D. x H: 60 x 13.5 mm, 13.5 mm Insertion Depth
LDPE
For use as plugs in bores or internal thread, or as caps for external thread
100 Pack
60 mm
Tapered Closure