Learn More
Poeppelmann™ KAPSTO™ GPN 300 F Closures
Offering multifunctional use for plugging internal threads and bore holes. Poeppelmann™ KAPSTO™ GPN 300 F Closures are manufactured from natural-colored PE-LD plastic.
Brand:  Poeppelmann™ 30001410000
 View more versions of this product
Product Code. 10695305
  • € 13.60 / Pack of 100
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Description
  • Flat-headed design
  • Firm fit due to crowned shank
  • Production in acc. to VDA Band 19 and ISO 16232
Specifications
Specifications
22 mm
For internal threads and bores
16.5 mm
Flat Top Closure Plug
7.5 mm