Learn More
Poeppelmann™ KAPSTO™ GPN 300 F Closures
Offering multifunctional use for plugging internal threads and bore holes. Poeppelmann™ KAPSTO™ GPN 300 F Closures are manufactured from natural-colored PE-LD plastic.
Brand:  Poeppelmann™ 30000100000
 View more versions of this product
Product Code. 10615865
  • € 11.70 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Description
  • Flat-headed design
  • Firm fit due to crowned shank
  • Production in acc. to VDA Band 19 and ISO 16232
Specifications
Specifications
20 mm
For internal threads and bores
13.7 mm
Flat Top Closure Plug
10 mm