Learn More
Poeppelmann™ KAPSTO™ GPN 300 F Closures
Offering multifunctional use for plugging internal threads and bore holes. Poeppelmann™ KAPSTO™ GPN 300 F Closures are manufactured from natural-colored PE-LD plastic.
Brand:  Poeppelmann™ 30000150000
 View more versions of this product
Product Code. 10587404
  • € 14.25 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Description
  • Flat-headed design
  • Firm fit due to crowned shank
  • Production in acc. to VDA Band 19 and ISO 16232
Specifications
Specifications
22 mm
For internal threads and bores
17.1 mm
Flat Top Closure Plug
10.5 mm