missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Inoculating L-Shaped Spreaders

Product Code. 12632285
Click to view available options
Quantity:
Pack of 1000
Quantity per Carton : 2000
Quantity per Carton : 6000
Subpacks of 5
Unit Size:
Pack of 1000
Pack of 2000
Pack of 400
Pack of 4000
Pack of 5000
Pack of 6000
This item is not returnable. View return policy

Product Code. 12632285

Brand: Gosselin™ ETAR06

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For spreading over the surface of solid media in microbiological applications. Gosselin™ Inoculating L-Shaped Spreaders are manufactured from polystyrene.

  • Curved end
  • Sterile and disposable
  • Packed in tamper-evident ZIP bags
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Description Individually Wrapped
Certifications/Compliance EC N°1935/2004, ISO 11137
Color White
Length (Metric) 148 mm
Quantity Quantity per Carton : 2000
Shape L-Shape
Material Polystyrene
Product Type Inoculating Spreader
Sterility Sterile
For Use With (Application) For Dairy, Environmental, Industrial, Microbiological Uses
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.