Learn More
Gosselin™ Inoculating L-Shaped Spreaders
Click to view available options
Quantity:
Pack of 1000
Quantity per Carton : 2000
Quantity per Carton : 6000
Subpacks of 5
Unit Size:
Pack of 1000
Pack of 2000
Pack of 400
Pack of 4000
Pack of 5000
Pack of 6000
Description
- Curved end
- Sterile and disposable
- Packed in tamper-evident ZIP bags
Specifications
Specifications
| Disposable | Single-use |
| Certifications/Compliance | EC N°1935/2004, ISO 11137 |
| Color | White |
| Length (Metric) | 148 mm |
| Quantity | Quantity per Carton : 6000 |
| Shape | L-Shape |
| Material | Polystyrene |
| Product Type | Inoculating Spreader |
| Sterility | Sterile |
| For Use With (Application) | For Dairy, Environmental, Industrial, Microbiological Uses |