missing translation for 'onlineSavingsMsg'
Learn More
Gold wire, 0.5mm (0.02in) dia, annealed, Premion™, 99.999% (metals basis), Thermo Scientific™ Product Code.: 15457425

Gold wire, 0.5mm (0.02in) dia, annealed, Premion™, 99.999% (metals basis), Thermo Scientific™

Product Code. 15457425
50cm
Click to view available options
Quantity:
10cm
50cm
250cm
This item is not returnable. View return policy

Product Code. 15457425

Brand: Thermo Scientific Alfa Aesar 047160.BW

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.967
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Quantity 50cm
Percent Purity 99.999%
Grade Premion™
Odor Odorless
Weight ≈3.79g/m
Assay (metals basis)
Chemical Name or Material Gold wire, 0.5mm (0.02 in.) dia., annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP17L005 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog Number047160.CB,047160.BS,047160.BW
Lot NumberP03L009 Certificate TypeCertificate of Analysis Date06/03/2025 Catalog Number047160.CB,047160.BS,047160.BW
Lot NumberM22L018 Certificate TypeCertificate of Analysis Date24/01/2025 Catalog Number047160.CB,047160.BS,047160.BW
Lot NumberY11H017 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number047160.CB,047160.BS,047160.BW
Lot NumberS28H021 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number047160.CB,047160.BS,047160.BW

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold wire, 0.5mm (0.02in) dia, annealed, Premion™, 99.999% (metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.