missing translation for 'onlineSavingsMsg'
Learn More

Gold rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)

Product Code. 11367397
Click to view available options
Quantity:
50 mm
Unit Size:
50mm
This item is not returnable. View return policy

Product Code. 11367397

Brand: Thermo Scientific Alfa Aesar 012555.DJ

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7440-57-5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-57-5
Quantity 50 mm
Percent Purity 99.99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.