Learn More
Gold Platinum Palladium powder, APS <5 micron, 99.9% (metal s basis), Thermo Scientific Chemicals
€ 282.00 - € 729.00
Specifications
Chemical Name or Material | Gold Platinum Palladium powder |
---|---|
Avg. Mol. Wt. or Mol. Wt. Range | Au:Pt:Pd; 40:40:20 wt% |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD00192403 |
UN Number | UN3089 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433820
|
Thermo Scientific Alfa Aesar
089726.03 |
1 g |
€ 282.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443820
|
Thermo Scientific Alfa Aesar
089726.06 |
5 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gold Platinum Palladium powder | |
(metals basis) | |
UN3089 | |
APS <5 Micron |
Au:Pt:Pd; 40:40:20 wt% | |
MFCD00192403 | |
99.9% | |
Powder |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only