missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold for AAS 1g/L HCl matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
€ 101.00 - € 251.00
Specifications
Concentration | 1g/L |
---|---|
Solution Contains | HCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12913125
|
Fisher Chemical
B 004 100 |
100 mL |
€ 101.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12923125
|
Fisher Chemical
B 004 500 |
500 mL |
€ 251.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
1g/L | |
HCl |