Learn More
PPP1R15A, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PPP1R15A.
Brand: Abnova PAB6009.100ug
Description
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The induction of this gene by ionizing radiation occurs in certain cell lines regardless of p53 status, and its protein response is correlated with apoptosis following ionizing radiation. [provided by RefSeq
Sequence: C-AAALDLSGRRGSpecifications
PPP1R15A | |
Polyclonal | |
Unconjugated | |
ELISA (1:128000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
PPP1R15A | |
PPP1R15A | |
A synthetic peptide corresponding to C-terminus of human PPP1R15A. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PPP1R15A. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
GADD34 | |
Goat | |
Antigen affinity purification | |
RUO | |
23645 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only