missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ GM 6001, Negative Control
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 1 mg |
| Description | Off-white |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | A cell impermeant negative control for the MMP inhibitor GM 6001 |
| Molecular Formula | C23H34N4O4 |
| Formula Weight | 430.54 |
| Synonym | (N-t-Butoxycarbonyl-L-leucyl-L-tryptophan methylamide) |
| Sensitivity | Moisture Sensitive |
| Solubility | Soluble in DMSO |
| Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction