missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Electrophoresis Grade, MP Biomedicals™

Product Code. p-26286 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11481642

Brand: MP Biomedicals 04808822

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycine, Electrophoresis Grade is for use in electrophoresis buffers and SDS-PAGE.

  • Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis.
  • Glycine is also a component of Towbin′s transfer buffer for Western blots.
  • It is an inhibitory neurotransmitter in the spinal cord and an allosteric regulator of NMDA receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 451°F
pH 4 (0.2 molar aq. soln.)
Specific Rotation ≤0.0065%
Solubility Information Soluble in water. Slightly soluble in alcohol and ethanol (95%). Almost insoluble in ether.
Density 1.1607 g/mL
Quantity 1 kg
Optical Rotation ≤0.0065%
Percent Purity ≥99.5%
Grade Electrophoresis
Physical Form Powder
pKa 2.35 at 25°C
Chemical Name or Material Glycine
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.