missing translation for 'onlineSavingsMsg'
Learn More

Glutathione Reduced, Cell Culture Reagent, ≥98%, MP Biomedicals™

Product Code. 11435202 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 11435202

Brand: MP Biomedicals 0219467905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glutathione Reduced, Cell Culture Reagent, ≥98% is used for protein purification and elutes GST-tagged protein from glutathione agarose.

  • Glutathione suppresses human immunodeficiency virus expression in chronically infected monocytic cells.
  • It is a useful tripeptide involved in many aspects of metabolism, including transport of g-glutanyl amino acids and reductive cleavage of disulfide bonds.
  • Glutathione is an endogenous antioxidant that plays a major role in reducing reactive oxygen species formed during cellular metabolism and the respiratory burst.
  • Glutathione-S-transferase catalyzes the formation of glutathione thioethers with xenobiotics, leukotrienes, and other molecules that have an electrophilic center.
  • Glutathione also forms disulfide bonds with cysteine residues in proteins.
  • Via these mechanisms, it can have the paradoxical effect of reducing the efficacy of anti-cancer agents.
  • Typical Working Concentration: May be used at 5-10 mM to elute glutathione S-transferase (GST) from glutathione agarose.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Especificaciones

Melting Point 195°C
Color White
Assay Percent Range >98%
Quantity 5 g
Solubility Information 292.5 mg/mL
Formula Weight 307.32
Percent Purity ≥98%
Grade Cell Culture
Physical Form Powder
Chemical Name or Material Glutathione Reduced, Cell Culture Reagent

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado