Learn More
Glassy carbon rod, 7mm (0.28in) dia, type 1, Thermo Scientific Chemicals
Used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used as electrical conductor ion-selective membrane.
€ 293.00 - € 796.00
Specifications
Chemical Name or Material | Glassy carbon rod |
---|---|
Packaging | Tube |
MDL Number | MFCD00133992 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384337
|
Thermo Scientific Alfa Aesar
038012.DJ |
50 mm |
€ 293.00
50mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394337
|
Thermo Scientific Alfa Aesar
038012.DM |
100 mm |
€ 476.00
100mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15451477
|
Thermo Scientific Alfa Aesar
038012.DR |
200 mm |
€ 796.00
200mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glassy carbon rod is used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used as electrical conductor ion-selective membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon rod | |
Tube | |
MFCD00133992 | |
Insoluble in water. |
Safety and Handling
missing translation for 'einecsNumber' : 231-153-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only