missing translation for 'onlineSavingsMsg'
Learn More

Glassy carbon rod, 5mm (0.2in) dia, type 2, Thermo Scientific Chemicals

Product Code. 11354307
Click to view available options
Quantity:
50 mm
100 mm
200 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Widely used as an electrode material in electrochemistry, for high temperature crucibles and as a component of some prosthetic devices.

Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a component of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
MDL Number MFCD00133992

Specifications

Packaging Tube
Quantity 50 mm
Solubility Information Insoluble in almost all solvents.
Chemical Name or Material Glassy carbon rod

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'