Learn More
Hirschmann™ Class A Volumetric Flasks with Hollow Glass Stopper
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
19/32 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
34/35 NS Standard Ground
45/40 NS Standard Ground
7/16 NS Standard Ground
Förpackningsstorlek
Each
Pack of 2
Beskrivelse
- With NS ground, hollow glass stopper
- With blue stain graduation
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Tekniske data
Tekniske data
| Certifications/Compliance | DIN EN ISO 1042 |
| Closure Material | Glass |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | 300 mm |
| Limit of Error | ±0.6 mL |
| Standard Taper Joint | 19/32 NS Standard Ground |
| Tolerance | ±0.6 mL |
| Graduated | Yes |
| Class | A |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion