Learn More
Kimble Chase™ Glass Tube for Potter-Elvehjem™ Tissue Grinder
Product Code. p-8003970
Click to view available options
Capacity (Metric):
0.5 mL
3 mL
8 mL
17 mL
45 mL
Size (Numerical):
18
20
21
22
23
24
Unit Size:
Each
Product Code. 10652473
Brand: Kimble Chase™ 8857520022
Only null left
Add to basket
Add to basket
Replacement Tube for Kimble™ Potter-Elvehjem™ Tissue Grinder.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 8 mL
Material Glass, PTFE
Product Type Tube
For Use With (Equipment) For Potter-Elvehjem Tissue Grinder
Size (Numerical) 22
Pestle Material PTFE