Learn More
Hirschmann™ Hollow Glass Stoppers
Product Code. 30090416
Click to view available options
Standard Taper Joint:
NS 10/19
NS 12/21
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Product Code. 30090416
Brand: Hirschmann™ 8900352
Only null left
Add to basket
Add to basket
For the safe and secure closure of flasks, bottles, tubing and other laboratury apparatus. Hirschmann Instruments™ Hollow Glass Stoppers are manufactured from clear glass.
  • Acc. to DIN 12 252
  • Available to fit a range of sizes
Specifications
For Use With (Equipment) For tight sealing of standard ground vessels
Quantity 1/Ea.
Standard Taper Joint NS 7/16
Material Glass
Product Type Hollow Stopper