Learn More
Rogo Sampaic™ Glass Staining Jars
Click to view available options
Slide Count:
10
20
Unit Size:
Each
Description
- With lid
- With removable rack
- Capacity of 8 to 20 slides, depending on size
Specifications
Specifications
| Material | Glass |
| Type | Staining Jar BPL010 |
| Slide Count | 20 |
| Dimensions (L x W) | 105 x 85 mm |
| Length (Metric) | 105 mm |
| Width (Metric) | 85 mm |
| Depth (Metric) | 75 mm |