Learn More
Hach Lange™ Glass Round Cuvettes
Made with glass, these 7 mL round cuvettes with rubber caps are exemplary water quality testing supplies.
Brand:  Hach Lange™ LCW906
Product Code. 10440874
  • € 78.90 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Made with glass material
  • 13mm round optical path length
  • 25 pieces, 7 mL, with rubber caps
  • 190 nm – 1100 nm wavelength range
Order Info
dimProductType
Specifications
Specifications
7 mL
25 Pack
Round Cuvette