Learn More
Hanna Instruments™ Glass Refillable Combination pH Electrode
Brand:  Hanna Instruments™ HI1053B
Product Code. 10385621
  • € 433.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Glass
0 to 12
-5°C to +100°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers