Learn More
Karl Hecht™ Glass Pasteur Pipettes
Click to view available options
Length (Metric):
150 mm
230 mm
Unit Size:
Pack of 250
Description
- Disposable
- Acc. to ISO 7712
- Manufactured of soda glass
- Long tapered tip
- With constriction in the upper part for cotton plug
- Capillary tip inside diameter of approx. 1mm, body approx. 90 x 7mm
- Short form and long form available
- 250 pieces/box, 1000 pieces/carton
- With CE marking
- 100 pieces/tube; 10 tubes/box
Specifications
Specifications
| Diameter (Metric) Outer | 7 mm |
| Material | Soda Lime Glass |
| For Use With (Application) | Pipetting |
| Quantity | 250/Box, 1000/Carton |
| Tip Style | Long Tapered |
| Length (Metric) | 230 mm |
| Disposable | Single-use |