Learn More
Rogo Sampaic™ Glass Hellendahl Staining Jar
Click to view available options
Slide Count:
8
Unit Size:
Each
Description
- Clear glass
- High wall shape
Specifications
Specifications
| Material | Glass |
| Type | Staining Jar |
| Slide Count | 8 |
| Dimensions (L x W) | 60 x 60 mm |
| Length (Metric) | 60 mm |
| Width (Metric) | 60 mm |
| Depth (Metric) | 100 mm |
Product Content Correction
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion