Learn More
Merck Glass Funnel for Vacuum Filtration
Shop All Merck Products
Click to view available options
Capacity (Metric):
300 mL
1 L
Description:
Ground Glass Seal
PTFE-faced
Unit Size:
Each
Description
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Specifications
Specifications
| Diameter (Metric)Top | 47 mm |
| Capacity (Metric) | 1 L |
| Material | Borosilicate Glass |
| Color | Clear |
| Description | Ground Glass Seal |