Learn More
WTW™ Glass Flow Through Vessel
Flow vessels for WTW conductivity measuring cells are perfectly designed to perform measurements without the interference of carbon dioxide through the air or to examine the conductivity of solutions in closed circuits.The overpressure in the system must not exceed 0.5 bar.
Brand: WTW™ 302730
Specifications
Flow-through Vessel |