Learn More
Eppendorf™ Glass Cuvette
Designed for a variety of applications
Brand: Eppendorf™ 4011100002
103.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Optical glass composition
- Available in a variety of models and sizes
Specifications
Cuvette | |
Glass | |
Spectrophotometer | |
Spectroscopy |
Model: 4011, semi-micro | |
1 Each | |
10 mm |
Product Content Correction
Product Title
Eppendorf™ Glass Cuvette > Model: 4011