Learn More
Hirschmann™ Glass covers
Click to view available options
Dimensions (L x W):
21 x 26 mm
24 x 40 mm
24 x 60 mm
Unit Size:
Pack of 1000
Description
- LOT number on packaging
- CE pursuant to IVD 98/79
- DIN 58 884
Specifications
Specifications
| Thickness (Metric) | #1 (0.13 to 0.16 mm) |
| Dimensions (L x W) | 24 x 40 mm |