Learn More
Hirschmann™ ringcaps™ Disposable Capillary Pipettes
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
Brand: Hirschmann™ 9600110
Description
- With ringmark
- Conformity certified
- Manufactured from durable DURAN™ glass
- Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
0.25% | |
Glass | |
10 μL | |
Single-use |
ISO 7550, CE, IVD 98/79 EC | |
Ring Mark | |
Capillary Pipette |