Learn More
Gilson™ Safe Aspiration Station
Product Code. p-4330379 Shop All Gilson Products
Click to view available options
Product Type:
Tubing Connector
Unit Size:
Each
Product Code. 11307830
Brand: Gilson™ F1077418
Vacuum-Controlled Safe Aspiration Station with small footprint.
  • Closed, stand-alone unit designed to prevent contamination hazards
  • Compact design with adjustable speed allowing quick aspiration
  • Always ready to use by simply pressing the handle button
  • Handle accommodates different types of adaptors to aspirate from a variety of vessels
  • Convenient coupling for quick bottle emptying
  • In-line hydrophobic filter traps aerosols
  • Connect two handles to the station with the Y-shape connector
  • Motor starts only when the vacuum pressure drops
  • Level detection prevents overflow
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tubing Connector
Color Clear
For Use With (Equipment) Gilson safe aspiration station
Quantity 1 Ea.