Learn More
Aalborg™ TIO-GFC Mounting Kit
Designed especially for the laboratory these kits offer a diverse economical way of acquiring flow measurement capabilities. Aalborg™ TIO-GFC Mounting Kit allows for easy changing of flow tubes in the field.
Brand: Aalborg™ KIT-TC-230EU-2C
This item is not returnable.
View return policy
Description
- Kits are shipped in convenient molded plastic carrying cases.
- Flow tubes and floats are interchangeable in frames supplied facilitating overlapping flow ranges.
- For non-corrosive fluids use the Aluminum Kit.
- For corrosives consider the Stainless Steel Kit.
- For corrosive applications at lower pressure or for high purity fluid service specify the PTFE Kit.
- Supplied with glass floats installed.
Specifications
230 V | |
GFC Mounting Kit |