Learn More
Aalborg™ TIO-GFC Mounting Kit
Designed especially for the laboratory these kits offer a diverse economical way of acquiring flow measurement capabilities. Aalborg™ TIO-GFC Mounting Kit allows for easy changing of flow tubes in the field.
Brand:  Aalborg™ KIT-TC-230EU-2C
Product Code. 12074029
  • € 146.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Kits are shipped in convenient molded plastic carrying cases.
  • Flow tubes and floats are interchangeable in frames supplied facilitating overlapping flow ranges.
  • For non-corrosive fluids use the Aluminum Kit.
  • For corrosives consider the Stainless Steel Kit.
  • For corrosive applications at lower pressure or for high purity fluid service specify the PTFE Kit.
  • Supplied with glass floats installed.
Specifications
Specifications
230 V
GFC Mounting Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers