Learn More
Amarell™ Standard Density Hydrometer without Thermometer, measuring span 0.200g/cm3
Calibrated for reference temperature 20°C. The standard density hydrometer without thermometer can be used for many general specific gravity measurements.
Brand:  Amarell™ H801372
 View more versions of this product
Product Code. 10129871
Description
Description
  • Reference temperature 20°C
  • Each hydrometer length is 280mm
Specifications
Specifications
1Each
±1g/cm3
280 mm
20°C
Density Hydrometer
0.002 g/cm3
1.800 to 2.000 g/cm3
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Amarell™ Standard Density Hydrometer without Thermometer, measuring span 0.200g/cm3 > Range: 1.800 to 2.000g/cm3; Length: 280mm; Accuracy: ±1g/cm3