Learn More
Thermo Scientific™ GeneBLAzer™ M1-NFAT-bla Jurkat Cells
GeneBLAzer™ M1-NFAT-bla Jurkat cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla Jurkat cell line.
Brand:  Thermo Scientific™ K1710
Product Code. 10656414
  • € 16300.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Description
GeneBLAzer™ M1-NFAT-bla Jurkat cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla Jurkat cell line. CellSensor™ NFAT-bla Jurkat cells (Cat. No. K1077) contain a beta-lactamase (bla) reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
M1-NFAT-bla Jurkat cells are functionally validated for Z'-factor and EC50 concentrations of carbachol. In addition, M1-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
In the brain, M1 activation mediates 'slow' neuronal excitability. Cortical and hippocampal muscarinic receptors are thought to be important in the attentional aspects of cognition. The predominant receptor subtypes in these brain areas are M1, M3, and M4. Therefore, M1 is a potential target for cognition, Alzheimer's, dementia, and schizophrenia. Studies on knock-out mouse models of M1 are also beginning to reveal potential functions of the receptor. Additional information on the muscarinic receptors can be found in reviews.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_000738
Dividing Cells
GeneBLAzer™ M1-NFAT-bla Jurkat Cells, 5 x 106 Cells, ∽5,000,000 cells/vial at 5,000,000 cells/mL Quantity of Cells, Jurkat Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
FRET
GeneBLAzer™ M1-NFAT-bla Jurkat cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: GeneBLAzer™ M1-NFAT-bla Jurkat cells, GeneBLAzer™ M1-NFAT-bla Jurkat Assay Protocol, Certificate of Analysis
GeneBLAzer™
BLA (Beta-Lactamase)
M1
Jurkat
GPCR
Cell-based beta lactamase reporter gene
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ M1-NFAT-bla Jurkat cells
• GeneBLAzer™ M1-NFAT-bla Jurkat Assay Protocol
• Certificate of Analysis
The GeneBLAzer™ M1-NFAT-bla Jurkat cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Fluorescence
M1
Single Cell Clone Origin
5 x 106 Cells
Jurkat
Cell Line
GeneBLAzer™