Learn More
Thermo Scientific™ GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 Cells
Description
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells contain the human Dopamine Receptor 2 (D2) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells (Cat. No. K1536) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Chimeric G protein, Gqo5.
The GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of apomorphine. In addition, GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
The D2 receptor is found primarily in the brain. There are two forms of D2 which are created by alternate splicing: a short form and a long form. This assay uses the long form, although little difference in ligand binding or signaling has been seen between the two forms.
Specifications
Specifications
| Accession Number | NM_000795 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | LiveBLAzer™-FRET B/G Loading Kit, Solution D, and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells • GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells should be stored in liquid nitrogen immediately upon receipt. |
| Description | GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 Cells, 2 x 106 Cells, ∽2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage |
| Detection Method | Fluorescence |
| For Use With (Application) | FRET |
| Gene Symbol | D2 |
| Includes | GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 cells, GeneBLAzer™ D2-Gqo5-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis |
| Population Status | Single Cell Clone Origin |
| Show More |