Learn More
Thermo Scientific™ GeneBLAzer™ CCKAR-NFAT-bla HEK 293T Cells
The GeneBLAzer™ CCKAR-NFAT-bla CHO-K1 cells contain the human Cholecystokinin A Receptor (CCKAR) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1548
Description
The GeneBLAzer™ CCKAR-NFAT-bla CHO-K1 cells contain the human Cholecystokinin A Receptor (CCKAR) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line. GeneBLAzer™ NFAT-bla CHO-K1 (Cat. No. K1447) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element stably integrated into CHO-K1 cells.
The CCKAR-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations using established ligands. In addition, CCKAR-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
Specifications
NM_000730.2 | |
Dividing Cells | |
Fluorescence | |
CCKAR | |
Single Cell Clone Origin | |
2 x 106 Cells | |
CCKAR | |
293T (HEK) | |
GPCR |
Cell-based beta lactamase reporter gene | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: • GeneBLAzer™ CCKAR-NFAT-bla HEK 293T cells • GeneBLAzer™ CCKAR-NFAT-bla HEK 293T Assay Protocol • Certificate of Analysis The GeneBLAzer™ CCKAR-NFAT-bla HEK 293T cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
GPCRs | |
CCKAR-NFAT-bla HEK 293T cells, CCKAR-NFAT-bla HEK 293T Assay Protocol, Certificate of Analysis | |
GeneBLAzer™ | |
BLA (Beta-Lactamase) | |
Dividing Cells | |
GeneBLAzer™ |