Läs mer
Läs mer
Agilent BioTek Gen6 Data Analysis Software
Handla allt< titel> Produkter
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Microplate Reader
Förpackningsstorlek
Each
Beskrivning
- Walk-up usability
- Both new and experienced users will be able to walk up and use Gen6 without extensive training required.
- Set up assay, analyze results, and export data in minutes
- With Gen6, users can set up their plate, read, and analyze results in minutes.
- Prebuilt experiment templates in Gen6
- Prebuilt experiment templates allow new users to quickly and easily get started with common assays and applications within Gen6.
- Advanced read functionality including kinetics and area scans
- Gen6 contains advanced functionality for kinetic assays. Users can easily set up time-course experiments and visualize data in real-time as the microplate reader collects it. Raw and calculated data can easily be exported with one-click into Microsoft Excel.
- Advanced reading modes, such as area scans, allow users to measure discrete points across a well for heterogeneous samples.
- Instrument compatibility: Synergy Neo2, Synergy H1, Synergy HTX, Synergy LX, Epoch 2, Epoch, and 800 TS
Specifikationer
Specifikationer
| For Use With (Equipment) | Microplate Reader |
| Type | Software |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering