Learn More
Grant Instruments Gel Card Rotor for LMC-56
Rotor for 24 gel-cards.
Brand: Grant Instruments Ri-24GC
Description
- Rotor for 24 Gel Cards for blood group serology testing (Forward Group, Reverse Group, RhD Type and 3 cell antibody screen).
- Compatible with LMC-56

Specifications
Stainless Steel | |
24 Gel Cards | |
1,500 rpm | |
1 kg | |
200 mm | |
2 Years | |
Autoclavable |
Swing-out Rotor | |
280 x g | |
100 mm | |
LMC-56 | |
24 | |
No | |
1 Ea. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.