Learn More
Trajan™ BP624 GC Capillary Columns
Click to view available options
Length (Metric):
30 m
30 mm
50 m
60 m
Diameter (Metric) Inner:
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
1.4 μm
1.8 μm
3 μm
1.2 mm
3 mm
Unit Size:
Each
Description
- Polar phase
- EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
- Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
Specifications
| Max. Temperature | 230/240°C |
| Diameter (Metric) Inner | 0.32 mm |
| Film Thickness | 1.8 μm |
| Length (Metric) | 30 m |