Learn More
Thermo Scientific™ Cover for Precision™ Water Baths
Product Code. 15365887 Shop All Thermo Scientific Products
Click to view available options
Type:
Gable Cover
Large Gassing Hood
Small Gassing Hood
Material:
Polycarbonate
Stainless Steel
For Use With (Equipment):
Precision 10L Water Baths
Precision 15L Shaking Water Baths
Precision 20L and 28L Water Baths
Precision 2L Water Bath
Precision 2L and 5L Water Baths
Unit Size:
Each
Product Code. 15365887
Brand: Thermo Scientific™ TSGPACL10
Only null left
Add to basket
Add to basket
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
Gable Covers
  • Enhanced hinged design supports open lid configuration and easy lid removal
  • Accommodate taller labware with extended height
  • Polycarbonate and stainless steel models available
  • Warranty varies by country, please contact us for details
Warranty
1 year
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 10 L
For Use With (Equipment) Precision 10L Water Baths
Type Gable Cover
Material Polycarbonate